Learn More
Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer.
Brand: Thermo Scientific™ 4000708
6310.90 NOK valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Description
More flexibility for your controlled-rate freezing applications with Thermo Scientific™ Racks for the CryoMed™ Controlled-Rate Freezer. Includes an assortment of cane, canister, straw and vial racks.
- Warranty varies by country, please contact us for details
Specifications
Silver | |
CryoMed 7452, 7453, 7473, 7454, 7455, 7474, 7458 | |
Vial Rack Holder |
Three 4 mL/5 mL Cryo Racks (Large) | |
Three 4 mL/5 mL Cryo Racks (Large) |
Product Suggestions
Customers who viewed this item also viewed
Special Offers
- Multiple Manufacturers