Learn More
Spectrum Labs™ Rack Set for CF-2 Fraction Collector
Spectrum Labs™ Rack Set for CF-2 Fraction Collector is a full set of racks for one specifically sized tube designed for use with the versatile CF-2 collector.
Brand:  Spectrum Labs™ 124856
 View more versions of this product
Product Code. 11969578
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Designed for use with the versatile CF-2 Fraction Collector
  • Complete set of tube racks designed to accommodate the tubes chosen
Specifications
Specifications
Fraction Collector Tube Rack
For up to 116 x 17 mm or 18 mm diameter tubes
CF-2 Fraction Collector
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Spectrum Labs™ Rack Set for CF-2 Fraction Collector > For up to 116 x 17mm or 18mm diameter tubes