missing translation for 'onlineSavingsMsg'
Learn More

Rabbit anti-Mouse IgG (H+L) Secondary Antibody, Invitrogen™

Rabbit Polyclonal Secondary Antibody

Brand:  Invitrogen A16162

Product Code. 15353956

  • 1010.00 NOK / 2mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The sensitivity of each lot of antibody is confirmed using ELISA. The specificity of each lot of antibody is confirmed by immunoelectrophoresis (IEP).

Mouse IgG (H+L) Polyclonal antibody specifically detects Mouse IgG (H+L) in Mouse samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mouse IgG (H+L)
Polyclonal
Unconjugated
Rabbit
Affinity chromatography
RUO
Mouse
Liquid
ELISA
> 4.5 mg/mL
PBS with 0.05% sodium azide; pH 7.2
Gamma Immunoglobins Heavy and Light chains
2 mg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.