missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAB27A Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17817-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene belongs to the small GTPase superfamily, Rab family. The protein is membrane-bound and may be involved in protein transport and small GTPase mediated signal transduction. Mutations in this gene are associated with Griscelli syndrome type 2. Alternative splicing occurs at this locus and four transcript variants encoding the same protein have been identified.Plays a role in cytotoxic granule exocytosis in lymphocytes. Required for both granule maturation and granule docking and priming at the immunologic synapse.Specifications
RAB27A | |
Polyclonal | |
Unconjugated | |
RAB27A | |
GS2, GTP binding protein Ram, HsT18676, Rab 27, RAB27, RAB27A, RAM, Ras related protein Rab 27A | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
5873 | |
-20°C | |
Liquid |
Immunoprecipitation, Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
0.33 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P51159 | |
RAB27A | |
RAB27A Fusion Protein Ag12136 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction