missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Recombinant Human Proprotein Convertase 1/PCSK1 Protein, CF
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Enzyme Activity
Brand: R&D Systems™ 2810-SE-010
This item is not returnable.
View return policy
Description
Measured by its ability to cleave the fluorogenic peptide substrate pERTKR-AMC (Catalog # ES013 ). The specific activity is >30 pmol/min/μg, as measured under the described conditions.Specifications
P29120 | |
Supplied as a 0.2 μm filtered solution in Tris, NaCl and Glycerol. | |
M.W. (Observed): 63-80 kDa, reducing conditions; M.W. (theoretical): 57 kDa | |
10 μg | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 6 months from date of receipt, -70°C as supplied. 3 months, -70°C under sterile conditions after opening. | |
<1.0 EU / 1 μg of the protein by the LAL method. | |
Unconjugated | |
>95%, by SDS-PAGE under reducing conditions and visualized by silver stain. |
Enzyme Activity | |
5122 | |
Proprotein Convertase 1/PCSK1 | |
Mouse myeloma cell line, NS0-derived human Proprotein Convertase 1/PCSK1 protein Ser111-Arg617, with a C-terminal 10-His tag | |
Mature form | |
PCSK1 | |
Recombinant | |
Proprotein Convertase 1/PCSK1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction