missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human FGFR3 (NP_001156685) VersaClone cDNA
Fibroblast Growth Factor Receptor 3
Brand: R&D Systems™
This item is not returnable.
View return policy
Description
The Human FGF R3 (NP_001156685) VersaClone cDNA clone contains the complete ORF for FGFR3, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The FGFR3 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.Order Info
Ships at ambient temperature.
Specifications
Gene ID (Entrez): 2261, 14184 | |
NP_001156685 | |
cDNA is provided in 10mM Tris-Cl, pH 8.5 | |
CD333, CEK, EC 2.7.10, FGF R3, fibroblast growth factor receptor 3, HSFGFR3EX, JTK4, JTK4thanatophoric dwarfism | |
Human, Mouse |
hFGFR3 | |
10 μg at 0.2 μg/μL | |
2440bp | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
For Research Use Only; Not for Use in Humans