missing translation for 'onlineSavingsMsg'
Learn More
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone, 99% Product Code.: 11354975

(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone, 99%

Product Code. 11354975
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11354975

Brand: Thermo Scientific Alfa Aesar L18358.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 66673-40-3 | C5H9NO2 | 115.132 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 66673-40-3
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.132
MDL Number MFCD00077791
InChI Key HOBJEFOCIRXQKH-SCSAIBSYSA-N
Synonym r---5-hydroxymethyl-2-pyrrolidinone, d-pyroglutaminol, 5r-5-hydroxymethyl pyrrolidin-2-one, r-5-hydroxymethyl pyrrolidin-2-one, r-5-hydroxymethyl-2-pyrrolidinone, r-5-hydroxymethyl-pyrrolidin-2-one, --d-pyroglutamol, r---5-hydroxymethylpyrrolidin-2-one, 5-hydroxymethyl-2-pyrrolidinoneShow MoreShow Less
PubChem CID 7128291
IUPAC Name (5R)-5-(hydroxymethyl)pyrrolidin-2-one
SMILES C1CC(=O)NC1CO

Specifications

Melting Point 83°C to 85°C
Boiling Point 147°C to 149°C (0.06 mmHg)
Quantity 1 g
Beilstein 5728422
Optical Rotation −30° (c=2 in Ethanol)
Formula Weight 115.13
Percent Purity 99%
Chemical Name or Material (R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZ28J010 Certificate TypeCertificate of Analysis Date26/01/2024 Catalog NumberL18358.06,L18358.03
Lot NumberZ21J011 Certificate TypeCertificate of Analysis Date10/01/2024 Catalog NumberL18358.06,L18358.03
Lot NumberY09H016 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL18358.06,L18358.03
Lot Number10137819 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL18358.06,L18358.03
Lot NumberF25S033 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL18358.06,L18358.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-(-)-5-(Hydroxymethyl)-2-pyrrolidinone, 99% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.