missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 95%, Thermo Scientific Chemicals

Product Code. p-4228336 Shop All Thermo Scientific Chemicals Products
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111769-27-8
Molecular Formula C11H17NO4S
Molecular Weight (g/mol) 259.32
MDL Number MFCD00040622,MFCD00040622,MFCD00066238
InChI Key BZXPLADBSZWDIH-UHFFFAOYNA-N
Synonym r-3-aminotetrahydrofuran tosylate, r-+-tetrahydro-3-furylamine p-toluenesulfonate salt, r---3-aminotetrahydrofuran toluene-4-sulfonate, r-+-3-aminotetrahydrofuran toluene-4-sulfonate, r---3-aminotetrahydrofurantoluene-4-sulfonate, 3-furanamine, tetrahydro-, r-, 4-methylbenzenesulfonate, c4h9no.c7h8o3s, r-tetrahydrofuran-3-amine-4-methylbenzenesulfonate, 3r-oxolan-3-amine; para-toluene sulfonate, 4-methylbenzenesulfonic acid; 3r-tetrahydrofuran-3-amine
PubChem CID 14243169
IUPAC Name 4-methylbenzenesulfonic acid;(3R)-oxolan-3-amine
SMILES NC1CCOC1.CC1=CC=C(C=C1)S(O)(=O)=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 137.3°C to 141°C
Color White
Elemental Analysis 48.91 to 52.99% C, 5.18 to 5.62% N
Infrared Spectrum Authentic
Assay Percent Range 95%
Quantity 1 g
Specific Rotation 4.3
Formula Weight 259.32
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material (R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt
Show More Show Less
compliance-icons
Product Identifier
  • (R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-(+)-3-Aminotetrahydrofuran p-toluenesulfonate salt, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.