missing translation for 'onlineSavingsMsg'
Learn More

(R)-2-Aminomethyl-1-Boc-pyrrolidine, 97%, Thermo Scientific Chemicals

381.03 NOK - 5880.00 NOK

Chemical Identifiers

CAS 259537-92-3
Molecular Formula C10H20N2O2
Molecular Weight (g/mol) 200.282
MDL Number MFCD03419256
InChI Key SOGXYCNKQQJEED-MRVPVSSYSA-N
Synonym r-2-aminomethyl-1-n-boc-pyrrolidine, r-1-boc-2-aminomethyl pyrrolidine, r-tert-butyl 2-aminomethyl pyrrolidine-1-carboxylate, tert-butyl 2r-2-aminomethyl pyrrolidine-1-carboxylate, r-2-aminomethyl-1-boc-pyrrolidine, r-2-aminomethyl-pyrrolidine-1-carboxylic acid tert-butyl ester, r-2-aminomethyl-1-tert-butoxycarbonyl pyrrolidine, tert-butyl r-2-aminomethyl-1-pyrrolidinecarboxylate, r-2-aminomethyl-1-n-boc-pyyrolidineShow MoreShow Less
PubChem CID 1512534
IUPAC Name tert-butyl (2R)-2-(aminomethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CN
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15459147
SDS View Documents
Thermo Scientific Alfa Aesar
H52419.MD
250 mg N/A N/A N/A
15439147
SDS View Documents
Thermo Scientific Alfa Aesar
H52419.03
1 g
1645.00 NOK
1g
Please to purchase this item. Need a web account? Register with us today!
15449147
SDS View Documents
Thermo Scientific Alfa Aesar
H52419.06
5 g
5880.00 NOK
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Reactant in the synthesis of diamino derivatives of [1, 2, 4] triazolo [1, 5-a][1, 3, 5] triazine(potent and selective adenosine A2A receptor antagonists).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reactant in the synthesis of diamino derivatives of [1, 2, 4] triazolo [1, 5-a][1, 3, 5] triazine(potent and selective adenosine A2A receptor antagonists).

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-2-Aminomethyl-1-Boc-pyrrolidine, 97%, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.