missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine, 97+%, Thermo Scientific Chemicals
886.50 NOK - 7968.00 NOK
Chemical Identifiers
CAS | 155806-35-2 |
---|---|
Molecular Formula | C38H50FeOP2 |
Molecular Weight (g/mol) | 640.61 |
MDL Number | MFCD00800284 |
InChI Key | YOYYLEHTACDQJL-BHACGSQPNA-N |
Synonym | 2r-1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino ferrocene, r,s-josiphos, r-s-ppf-p cy 2, r---1-s-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine, r---1-s-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine ethanol adduct, r-1-sp-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine, ferrocene, 1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino-, 2r, ferrocene,1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino-, 2r |
PubChem CID | 10984818 |
IUPAC Name | cyclopentane;dicyclohexyl-[(1R)-1-(2-diphenylphosphanylcyclopentyl)ethyl]phosphane;iron |
SMILES | [Fe].CCO.c1cccc1.C[C@@H](P(C1CCCCC1)C1CCCCC1)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | |||||
10670893
|
Thermo Scientific Acros
370671000 |
100 mg | Glass bottle |
886.50 NOK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10038981
|
Thermo Scientific Acros
370675000 |
500 mg | Glass bottle |
2927.57 NOK
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10094703
|
Thermo Scientific Acros
370670025 |
2.5 g | Glass bottle | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
155806-35-2 | |
640.61 | |
YOYYLEHTACDQJL-BHACGSQPNA-N | |
10984818 | |
[Fe].CCO.c1cccc1.C[C@@H](P(C1CCCCC1)C1CCCCC1)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1 |
C38H50FeOP2 | |
MFCD00800284 | |
2r-1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino ferrocene, r,s-josiphos, r-s-ppf-p cy 2, r---1-s-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine, r---1-s-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine ethanol adduct, r-1-sp-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine, ferrocene, 1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino-, 2r, ferrocene,1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino-, 2r | |
cyclopentane;dicyclohexyl-[(1R)-1-(2-diphenylphosphanylcyclopentyl)ethyl]phosphane;iron |
Specifications
155806-35-2 | |
100.0 | |
Authentic | |
C38H50FeOP2 | |
- 350.00 (20.00°C c=0.4,CHCl3) | |
2r-1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino ferrocene, r,s-josiphos, r-s-ppf-p cy 2, r---1-s-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine, r---1-s-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine ethanol adduct, r-1-sp-2-diphenylphosphino ferrocenyl ethyldicyclohexylphosphine, ferrocene, 1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino-, 2r, ferrocene,1-1r-1-dicyclohexylphosphino ethyl-2-diphenylphosphino-, 2r | |
[Fe].CCO.c1cccc1.C[C@@H](P(C1CCCCC1)C1CCCCC1)c1cccc1P(C1=CC=CC=C1)C1=CC=CC=C1 | |
640.61 | |
640.61 | |
Crystalline Powder or Crystals |
97.5 | |
White | |
97% min. (HPLC) | |
MFCD00800284 | |
− 350.00 | |
YOYYLEHTACDQJL-BHACGSQPNA-N | |
cyclopentane;dicyclohexyl-[(1R)-1-(2-diphenylphosphanylcyclopentyl)ethyl]phosphane;iron | |
10984818 | |
97+% | |
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine, 98% |
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
(R)-(-)-1-[(S)-2-(Diphenylphosphino)ferrocenyl]ethyldicyclohexylphosphine, 97+%, Thermo Scientific Chemicals