missing translation for 'onlineSavingsMsg'
Learn More

(R)-(+)-1,2-Epoxybutane, 98%, Thermo Scientific Chemicals

Product Code. 15404828
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 3760-95-0 | C4H8O | 72.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3760-95-0
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.107
MDL Number MFCD02683444
InChI Key RBACIKXCRWGCBB-SCSAIBSYSA-N
Synonym r-+-1,2-epoxybutane, r-2-ethyloxirane, 2r-2-ethyloxirane, r-1,2-epoxybutane, oxirane, ethyl-, 2r, r-epoxybutane, pubchem14098, r-+-ethyloxirane, r-+-butylene oxide
PubChem CID 11147670
IUPAC Name (2R)-2-ethyloxirane
SMILES CCC1CO1

Specifications

Density 0.83
Boiling Point 60°C to 64°C
Flash Point −12°C (10°F)
Refractive Index 1.381
Quantity 5 g
UN Number UN3022
Optical Rotation +10° (Neat)
Formula Weight 72.11
Percent Purity 98%
Chemical Name or Material (R)-(+)-1,2-Epoxybutane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-(+)-1,2-Epoxybutane, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.