missing translation for 'onlineSavingsMsg'
Learn More

(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine, ChiPros™ 99+%, ee 98+%, Thermo Scientific™

303.00 NOK - 2695.00 NOK

Chemical Identifiers

CAS 23357-46-2
Molecular Formula C10H13N
Molecular Weight (g/mol) 147.221
MDL Number MFCD00671629
InChI Key JRZGPXSSNPTNMA-SNVBAGLBSA-N
Synonym r-1,2,3,4-tetrahydronaphthalen-1-amine, r---1,2,3,4-tetrahydro-1-naphthylamine, 1r-1,2,3,4-tetrahydronaphthalen-1-amine, r-1,2,3,4-tetrahydro-1-naphthylamine, 1-naphthalenamine, 1,2,3,4-tetrahydro-, 1r, r-1-amino-1,2,3,4-tetrahydronaphthalene, r-1, 2, 3, 4-tetrahydro-1-naphthylamine, r-1-aminotetraline, tetralin-1alpha-amine, r-1-aminotetralinShow MoreShow Less
PubChem CID 7058072
IUPAC Name (1R)-1,2,3,4-tetrahydronaphthalen-1-amine
SMILES C1CC(C2=CC=CC=C2C1)N
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11313916
SDS View Documents
Thermo Scientific Alfa Aesar
L17021.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11323916
SDS View Documents
Thermo Scientific Alfa Aesar
L17021.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11333916
SDS View Documents
Thermo Scientific Alfa Aesar
L17021.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. Chiral amines play an important role in stereoselective organic synthesis. They are used directly as resolving agents, building blocks or chiral auxiliaries.

Solubility
Soluble in water (6.91g/L at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with oxidizing agents.
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'einecsNumber' : 000-000-0

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(R)-(-)-1,2,3,4-Tetrahydro-1-naphthylamine, ChiPros™ 99+%, ee 98+%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.