Learn More
PYREX™ Quickfit™ Steam Distillation Splash Head
Distillation head with steam connection
Brand: Quickfit SH16/19
6768.87 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- For use with rotary evaporators
- Prevents possible contamination of the condenser from 'bumping' solvents
- Features a coarse porosity 1 sinter to reduce carry over
- Splash heads feature a 29/32 socket
Specifications
Anti-splash with sinter | |
Splash head adapter | |
Rotary evaporators | |
1 Each | |
90 mL |
Borosilicate Glass | |
Prevents possible contamination of the condenser from 'bumping' solvents | |
Coarse, porosity 1 sinter | |
Cone: 19/26, Socket: 29/32 |
Product Content Correction
Product Title
PYREX™ Quickfit™ Steam Distillation Splash Head > Cone Shape: 19/26; For use with: Prevents possible contamination of the condenser from 'bumping' solvents