Learn More
Quickfit™ Right Angle Tubing Adapter
Quickfit™ Right Angle Tubing Adapter are available in a variety of cone sizes.
Brand: Quickfit MF10/4B/SC
326.63 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Quickfit™ Right Angle Tubing Adapter are available in a variety of cone sizes.
- With right angle connection for attachment of rubber hose etc
- Detachable plastic screwthread connectors allow flexible tubing to be fitted easily and safely
Specifications
29/32 | |
Detachable Plastic screw thread connector | |
Right angle tubing adapter |
Flexible tubing, Rubber hose | |
Borosilicate Glass |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
New Offer Save Now -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%