Learn More
Quickfit™ Reduction Socket Adapter
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
Brand:  Quickfit DA23
301.41 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 12634891
  • 455.00 NOK / Pack of 2
Estimated Shipment: 25-04-2025
to see stock.
Explore more special offers
Description
Made of borosilicate glass. Pyrex Labware™ Quickfit™ Reduction Socket Adapter connects large sized ground glass joints with smaller sized ones.
  • Useful for the connection of dissimilar sized ground glass joints
  • Small sized socket at the top, large sized cone at the bottom
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cone Size: 24/29
Small socket, large cone
Reduction adapter
Dissimilar sized ground Glass joint
Borosilicate Glass
19/26
Documents
One moment while we fetch your results.
Product Title
Quickfit™ Reduction Socket Adapter > Cone Size: 24/29; Socket Size: 19/26