Learn More
Quickfit™ Reaction Vessel Bubbler
Ideal for closing reaction mixture from the atmosphere.
Brand:  Quickfit GB1/24
 View more versions of this product
Product Code. 15768017
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Side arm to facilitate filling with mineral oil
  • Bubblers have capacity of approx 5ml
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Reaction Vessel Bubbler
24/29
Product Title
Quickfit™ Reaction Vessel Bubbler > Product type: Size: 24/29