Learn More
Quickfit™ Reaction Vessel Bubbler
Ideal for closing reaction mixture from the atmosphere.
Brand: Quickfit GB1/19
8503.19 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
- Side arm to facilitate filling with mineral oil
- Bubblers have capacity of approx 5ml
Specifications
Reaction Vessel Bubbler | |
Ideal for closing reaction mixture from the atmosphere |
Special Offers
-
Multiple Manufacturers
Price-Shattering Deals on Lab Glassware
Stock up on lab beakers, flasks and more before these deals disappear.Promo Code: 24123Expires: 2025-06-14
Product Content Correction
Product Title
Quickfit™ Reaction Vessel Bubbler > Cone Size: 19/26