Learn More
Pyrex™ Quickfit™ Metal Retaining Clips
Use with soxhlet extractors, flat flange lids and reaction vessels. Pyrex™ Quickfit™ Metal Retaining Clips apply positive pressure to both surfaces with spring wire design.
448.28 NOK - 512.00 NOK
Specifications
Material | Stainless Steel |
---|---|
For Use With (Application) | Securing flat flange lids to wide neck reaction flasks and culture vessels |
Description | Spring wire design |
Product Code | Brand | Size | Diameter (Metric) Inside | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Diameter (Metric) Inside | Price | Quantity & Availability | |||||
11312015
|
Pyrex™
JC100F |
FG100 | 100 mm |
512.00 NOK
Each |
Estimated Shipment: 28-07-2025
Log in to see stock. |
|||||
12405286
|
Pyrex™
JC50F |
FG50 | 50 mm | N/A | ||||||
10166304
|
Pyrex™
JC75F |
FG75 | 75 mm |
511.00 NOK
Each |
Estimated Shipment: 09-09-2025
Log in to see stock. |
|||||
Description
Use with soxhlet extractors, flat flange lids and reaction vessels. Pyrex™ Quickfit™ Metal Retaining Clips apply positive pressure to both surfaces with spring wire design.
- For securing flat flange lids to wide neck reaction flasks and culture vessels
- Stainless steel construction
Specifications
Stainless Steel | |
Securing flat flange lids to wide neck reaction flasks and culture vessels | |
Spring wire design |