Learn More
SVL™ Quick Release Clamp for SVL™ Flat-flange Joints
Easy to use metal clamps are designed to securely clamp SVL™ flange lids to SVL™ wide neck reaction vessels.
Brand:  SVL 295-02
1434.70 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 15707947
  • 2470.00 NOK / Each
Price-Shattering Deals on Lab Glassware
Use promo code 24123 at checkout
View Available Promotions
Only null left
Estimated Shipment: 29-08-2025
Description
  • One piece construction
  • Manufactured from stainless steel
  • Can be secured and removed in a single movement with an adjustable locking tab
TRUSTED_SUSTAINABILITY
Specifications
Specifications
1/Pk.
Stainless Steel
For Securing SVL Lids To Reaction Vessels
Quick Release Clamp
For 100mm Flange
Certificates
Promotions
Promotions
Product Content Correction
Product Title
SVL™ Quick Release Clamp for SVL™ Flat-flange Joints > Diameter: 14mm