Learn More
Quickfit™ Hexagonal Hollow-blown Glass Stopper
Ideally suited for use with volumetric flasks.
Brand:  Quickfit TRF698
966.08 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
 View more versions of this product
Product Code. 12481788
  • 1100.00 NOK / Pack of 5
Estimated Shipment: 15-08-2025
to see stock.
Explore more special offers
Description
Ideally suited for use with volumetric flasks.
  • Lower closed end of stopper features 'drip-tip' to ensure adhered liquid is returned to flask
  • Hexagonal head prevents stopper from rolling and provides good grip
  • Flat head allows stopper to be stood on end, thus preventing contamination of ground surface
  • Hollow-blown to reduce weight
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Volumetric flasks
34/35
Glass
5/Pk.
NS 34/35
Hollow Stopper
Product Title
Quickfit™ Hexagonal Hollow-blown Glass Stopper > Cone Size: NS 34/35