Learn More
Quickfit™ Three-Neck Round-Bottom Glass Flask
Useful for introducing gas inlet tubes and thermometers into the flask. The Quickfit Jointed Ware™ Quickfit™ Three-Neck Round-Bottom Flask has precision-ground sockets from borosilicate glass.
Brand:  Quickfit FR50/2S/11A
889.92 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 12413631
  • 1010.00 NOK / Each
Estimated Shipment: 04-08-2025
to see stock.
Explore more special offers
Description
Useful for introducing gas inlet tubes and thermometers into the flask. The Quickfit Jointed Ware™ Quickfit™ Three-Neck Round-Bottom Flask has precision-ground sockets from borosilicate glass.
  • With highest-quality Quickfit™ ground glass socket
  • Side neck useful for introducing gas inlet tubes and thermometers etc., into the flask
  • Manufactured from borosilicate glass for highest resistance to thermal shock and chemical attack
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ISO 383
Borosilicate Glass
19/26 (Centre), 14/23 (side)
50 mL
92 mm
Round
Documents
Product Information
Certificates
Special Offers
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    25
    Hours
    5
    Minutes
    20
    Seconds
    26
Product Content Correction
Product Title
Quickfit™ Three-Neck Round-Bottom Glass Flask > Capacity: 50mL; Socket: 19/26 (Centre), 14/23 (side)