Learn More
Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask
Use these flasks for a variety of applications. Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask are manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock.
Brand: Quickfit FR5/1S
925.38 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Description
Use these flasks for a variety of applications. Quickfit™ Borosilicate Glass Short Neck Round Bottom Flask are manufactured from borosilicate glass for corrosion-free performance and resistance to thermal shock.
- With highest quality Quickfit™ ground glass socket
- Manufactured from borosilicate glass for corrosion free performance and resistance to thermal shock
- Comprehensive range of 46 flasks from 5mL to 10,000ml
- Body diameter and wall thickness comply with relevant ISO 1773 and ISO 4797 requirements
Specifications
ISO 1773, ISO 4797 | |
Borosilicate Glass | |
Short | |
Round |
5 mL | |
65 mm | |
14/23 | |
25 mm |