Learn More
Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
Brand: Quickfit FF150/5S
This item is not returnable.
View return policy
Description
Manufactured from borosilicate glass. Quickfit Jointed Ware™ Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask complies with ISO 1773 and ISO 4797.
- Complies with ISO 1773 and ISO 4797
- With highest quality Quickfit™ ground glass socket
- Borosilicate glass for highest resistance to thermal shock and chemical corrosion
- Flat base allows use with hotplates
Specifications
ISO 1773, ISO 4797 | |
Borosilicate Glass | |
Short | |
Flat |
150 mL | |
115 mm | |
34/35 |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product information
File size: 2.8 MB
Product Content Correction
Product Title
Quickfit™ Borosilicate Glass Short Neck Flat Bottom Flask > Capacity: 150mL; Socket: 34/35