Learn More
Quickfit™ Borosilicate Glass Reversible Condenser Coil
Constructed of Pyrex™ Borosilicate Glass. Quickfit™ Borosilicate Glass Reversible Condenser Coil is fitted on each end with a different size cone to reduce the need for adapters. Includes detachable plastic connectors for the attachment of flexible tubing.
2370.00 NOK
Product Code | Brand | Standard Taper Joint No | Length (Metric) Jacket | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No | Length (Metric) Jacket | Price | Quantity & Availability | |||||
15547780
|
Quickfit
CX3/23/SC |
19/26 and 24/29 | 315 mm (Overall) 160 mm (Effective) |
2370.00 NOK
Pack of 2 |
Estimated Shipment: 22-08-2025
Log in to see stock. |
|||||
15557780
|
Quickfit
CX3/35/SC |
24/29 and 34/35 | 335 mm (Overall) 160 mm (Effective) |
2370.00 NOK
Pack of 2 |
Estimated Shipment: 02-10-2025
Log in to see stock. |
|||||
Description
- Highly versatile reflux condenser
- Each end of the condenser is fitted with a different size cone thus reducing the need for adapters
- With detachable plastic connectors for the attachment of flexible tubing
Specifications
Borosilicate Glass | |
Condenser Coil |
Product Title
Quickfit™ Borosilicate Glass Reversible Condenser Coil