Learn More
Quickfit™ Borosilicate Glass Iodine Flask
Complies with BS 2735
Brand: Quickfit FIB500/3
1619.75 NOK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Complies with BS 2735
- Complies with BS 2735
- For the determination of iodine
- Flask has ground socket and cup shaped top
- Stopper has extended handle which projects well above the liquid seal
Note: Stopper is not included; sold separately.
Specifications
BS 2735 | |
500 mL | |
24/29 |
Top style: Cup shaped | |
Borosilicate Glass |