Learn More
Quickfit™ Borosilicate Glass Drying Angled Tube
Keep material securely dry. QuickFit™ Borosilicate glass drying angled cone to fit QuickFitT™ apparatus.
402.00 NOK - 2870.00 NOK
Specifications
Material | Borosilicate Glass |
---|---|
Product Type | Tube |
For Use With (Equipment) | Quickfit™ apparatus |
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint No. | Price | Quantity & Availability | ||||||
12088371
|
Quickfit
MF23/1 |
14/23 |
402.00 NOK
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
||||||
12098371
|
Quickfit
MF23/2 |
19/26 |
458.00 NOK
Each |
Estimated Shipment: 03-09-2025
Log in to see stock. |
||||||
12008381
|
Quickfit
MF23/3 |
24/29 |
544.00 NOK
Each |
Estimated Shipment: 30-07-2025
Log in to see stock. |
||||||
15527940
|
Quickfit
MF23/4 |
29/32 |
2870.00 NOK
Pack of 5 |
Estimated Shipment: 22-07-2025
Log in to see stock. |
||||||
Description
- For use with a desiccant such as calcium chloride
- Ground glass cone to fit Quickfit™ apparatus
Specifications
Borosilicate Glass | |
Tube | |
Quickfit™ apparatus |
Product Title
Quickfit™ Borosilicate Glass Drying Angled Tube