Learn More
Quickfit™ Allihn Condenser with Ground Cone
Ideal for reflux applications.
Brand: Quickfit CX7/03/SC
1553.73 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
This item is not returnable.
View return policy
Description
Ideal for reflux applications.
- Inner bulb sections provide increased cooling surface
- With detachable plastic screwthread connectors for easy and safe attachment of flexible tubing
Specifications
Ideal for reflux work | |
405 mm | |
Borosilicate Glass |
405 mm | |
24/29 | |
Allihn Condenser |
Product information
File size: 2.8 MB
Product Content Correction
Product Title
Quickfit™ Allihn Condenser with Ground Cone > Cone Size: 24/29; Length: 405mm; Effective Length: 250mm