Learn More
Pyrex™ Quickfit™ Arsenic Determincation Apparatus
Constructed of borosilicate glass. Pyrex™ Arsenic Determination Apparatus provides a method of analysis for arsenic and antimony determination.
Brand:  Pyrex™ 1AD/1
2036.55 NOK valid until 2025-06-13
Use promo code "24123" to get your promotional price.
Product Code. 11991578
  • 2155.00 NOK / Each
Only null left
Estimated Shipment: 02-10-2025
Description
Constructed of borosilicate glass. Pyrex™ Arsenic Determination Apparatus provides a method of analysis for arsenic and antimony determination.
  • Designed for the estimation of arsenic and antimony
  • Method uses the formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 538nm or 5380Ă….
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Borosilicate Glass (3.3)
Formation of a complex between arsine and silver diethyldithiocarbamate with subsequent measurement at 538nm or 5380Ă….
1 Each
Arsenic Determination Apparatus
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Pyrex™ Quickfit™ Arsenic Determincation Apparatus > 100mL flask