missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Quickfit™ Angled Air/Stream Inlet Tube

Cut to length with glassblower. Pyrex™ Quickfit™ Angled Air/Steam Inlet Tube includes a 90° bend and detachable screwthread connector for easy flexible tubing insertion.

Brand:  Pyrex™ MF15/1B/SC

 View more versions of this product

Product Code. 11362045

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

Cut to length with glassblower. Pyrex™ Quickfit™ Angled Air/Steam Inlet Tube includes a 90° bend and detachable screwthread connector for easy flexible tubing insertion.

  • With detachable plastic screwthread connector to facilitate the attachment of 9mm bore flexible tubing
  • Ground glass cone on stem fits Quickfit™ sockets
  • Lower end of tube left plain for Glassblower to cut to length and draw to jet
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Angled Inlet Tube
150 mm
1 Each
14/23
Borosilicate Glass
9mm bore tubing
Detachable plastic screwthread connector
90°
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers