Learn More
SCAT Quick-Lock Connectors
Quick-Lock Connectors provide easy and safe connection of capillary tubing and SCAT Safety Caps. Connect and disconnect with a click - without dripping or air bubbles. The valve closes automatically when disconnected. Included PFA fittings allow all common hose diameters to be connected.
Brand:  SCAT 360183
836.96 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 17312860
  • 995.00 NOK / 1 set
Estimated Shipment: 13-05-2025
to see stock.
Explore more special offers
Description
The Quick-Lock Connector sets are suitable for connecting 2 capillaries (Set 1) or one capillary and one SCAT Safety Cap (Set 2).
  • Made of chemically resistant glass-filled polypropylene, PEEK™ materials
  • Leak protection thanks to integrated shut-off valves
  • Closed system for drip-free disconnection with very low air entrapment
  • Minimizes contamination and no need to purge air bubbles from the system
  • Suitable for capillaries with outer diameter 1.6 (1/16 in.) / 2.3 /3.2 (1/8 in.) mm
  • User-friendly, ergonomic design, enables safe connection
  • Up to vacuum 15 psi, 1.0 bar pressure resistance
Specifications
Specifications
HPLC Systems
Capillaries, Tubes and Safety Caps
360180 Quick-Lock Connector (m), 360179 Quick-Lock Connector (f)
Quick-Lock Connector Set I
Videos
  • HOW TO Use HPLC Quick Connectors with SCAT Safety Caps
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      HOW TO Use HPLC Quick Connectors with SCAT Safety Caps
    Product Title
    SCAT Quick-Lock Connectors > Type: Quick-Lock Connector Set I