Learn More
Invitrogen™ Qubit™ Flex Assay Tubes
Strip of eight tubes designed for the Qubit™ Flex.
Brand: Invitrogen™ Q33252
1503.40 NOK valid until 2025-07-04
Use promo code "24112" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
- Capacity: 200 ÎĽL
- Thin-walled
- Polypropylene tubes
- For use with the Qubit™ Flex Fluorometer
- Quantity: 125 tube strips
- Strips of eight tubes
Specifications
200 ÎĽL | |
125 tube strips | |
Qubit Flex Fluorometer | |
DNA, RNA or Protein Quantitation |