Learn More
Invitrogen™ Qubit™ Assay Reservoirs
Reservoirs that facilitate the repeated multi-channel of Qubit™ Reagents.
Brand: Invitrogen™ Q33253
1480.07 NOK valid until 2025-07-04
Use promo code "24112" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24112” to get your promotional price
Description
- Facilitates the repeated multi-channel pipetting of Qubit™ reagents into Qubit™ Flex Assay Tube Strips
- For use with Qubit™ Flex Fluorometer
Specifications
Qubit Flex Fluorometer | |
DNA, RNA or Protein Quantitation |
Promotions
-
Multiple Manufacturers
Cancer Research Campaign
Save big on essential products for cancer researchPromo Code: 24147Expires: 2025-07-31
-
Multiple Manufacturers
New Offer Save Now