Learn More
Hellma™ Quartz Flow Through Cells Far UV
9375.00 NOK - 13690.00 NOK
Product Code | Brand | Path Length | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Path Length | Price | Quantity & Availability | ||||||
12647201
|
Hellma™
170-000-2-40 |
2 mm |
9375.00 NOK
Each |
Estimated Shipment: 29-08-2025
Log in to see stock. |
||||||
11716274
|
Hellma™
170-700-2-40 |
2 mm |
10380.00 NOK
Each |
Estimated Shipment: 22-10-2025
Log in to see stock. |
||||||
11774296
|
Hellma™
170700-0.1-40 |
0.1 mm |
13690.00 NOK
Each |
Estimated Shipment: 04-07-2025
Log in to see stock. |
||||||
11967105
|
Hellma™
170-700-1-40 |
1 mm |
10155.00 NOK
Each |
Estimated Shipment: 18-09-2025
Log in to see stock. |
||||||
Specifications
Flow-through Cell Far UV | |
Quartz |
Product Title
Hellma™ Quartz Flow Through Cells Far UV