Learn More
Macherey-Nagel™ Quantofix™ Test Strips
Perform fast, easy analyses on the spot. Macherey-Nagel™ Quantofix™ Test Strips are ready to use semi-quantitative tests that change color depending on the concentration of the analyte in the sample. Read results using the color scale or with the Quantofix™ Relax.
Brand:  Macherey-Nagel™ 91330
 View more versions of this product
Product Code. 11306574
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Perform fast, easy analyses on the spot. Macherey-Nagel™ Quantofix™ Test Strips are ready to use semi-quantitative tests that change color depending on the concentration of the analyte in the sample. Read results using the color scale or with the Quantofix™ Relax.
Quantofix™ test strips meet all requirements of a modern rapid test. Analytical professionals as well as occasional testers appreciate the test strips for the fast and easy analysis directly at the point of interest. Often, these tests are used to quickly check whether important parameters are in the desired range. They deliver an immediate result and thus enable a fast response.
  • Rapid
    • Just Dip & Read
    • Results within seconds
    • Always ready for use
  • Easy
    • No calibration
    • No maintenance
    • No accessories
  • Reliable
    • Desiccant in the stopper for optimal protection of the strips against humidity
    • Color chart confirmed with traceable standards
    • Automatic evaluation with Quantofix™ Relax for safe documentation
Specifications
Specifications
White to Dark Red
Total Iron 1000
Semi Qualitative Test Strips
Determination of Total Iron 1000
5 - 1000mg/L Fe
30 sec.
30°C
Total Iron
2,2'-bipyridine
100 Pack
0, 5, 20, 50, 100, 250, 500, 1000mg/L Fe2+/3+
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Macherey-Nagel™ Quantofix™ Test Strips > Color Indicator: White to Dark Red