Learn More
Socorex™ Qualitix™ Sterilization Pouch
Fits any small to mid-size sterilization device and provides easy and effective wrapping of items. Made of robust medical grade paper, they tightly close with self-sealing strip to maintain sterility. Content remains visible through PET/CPP transparent laminated film.
Brand:  Socorex™ 3210.1936
 View more versions of this product
Product Code. 15740679
  • 692.00 NOK / Pack of 200
Only null left
Estimated Shipment: 04-07-2025
Description
  • Robust durable material
  • Quick content identification
  • Large self-sealing strip, easy to close
  • Chevron top seal for easy opening
  • Suitable for steam and EO gas sterilization
  • Two sterilization indicators
  • Five different sizes
  • Conformity to ISO 17665-06, ISO 11135, CE
Specifications
Specifications
Autoclavable
Self-adhesive
190 x 360 mm
ISO 17665-06, ISO 11135, CE
Paper
Videos
  • Socorex Qualitix sterilisation pouches
    Video Player is loading.
    Current Time 0:00
    Duration 0:38
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:38
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Socorex Qualitix sterilisation pouches
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Socorex™ Qualitix™ Sterilization Pouch > 190 x 360 mm