Learn More
QDPR Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14908-1-AP-150UL
 View more versions of this product
Product Code. 16833814
  • 4796.50 NOK / 150µL
Explore more special offers
Description
Description
This gene encodes the enzyme dihydropteridine reductase, which catalyzes the NADH-mediated reduction of quinonoid dihydrobiopterin. This enzyme is an essential component of the pterin-dependent aromatic amino acid hydroxylating systems. Mutations in this gene resulting in QDPR deficiency include aberrant splicing, amino acid substitutions, insertions, or premature terminations. Dihydropteridine reductase deficiency presents as atypical phenylketonuria due to insufficient production of biopterin, a cofactor for phenylalanine hydroxylase.
Specifications
Specifications
QDPR
Polyclonal
Unconjugated
QDPR
DHPR, Dihydropteridine reductase, HDHPR, PKU2, QDPR, SDR33C1
Rabbit
Antigen Affinity Chromatography
RUO
110391, 5860, 64192
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.21 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P09417, P11348, Q8BVI4
Qdpr
QDPR Fusion Protein Ag6705
150 μL
Primary
Human, Mouse, Rat