missing translation for 'onlineSavingsMsg'
Learn More
Pyrrolidine-3-carboxylic acid, 98% Product Code.: 15549473
compliance-icons

Pyrrolidine-3-carboxylic acid, 98%

Product Code. p-7077776 Shop All Thermo Scientific Chemicals Products
1 g, 1g
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15549473

Brand: Thermo Scientific Acros 459110010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 59378-87-9 | C5H9NO2 | 115.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59378-87-9
Molecular Formula C5H9NO2
Molecular Weight (g/mol) 115.13
InChI Key JAEIBKXSIXOLOL-UHFFFAOYSA-N
Synonym 3-pyrrolidinecarboxylic acid, 3-pyrrolidine carboxylic acid, beta-proline, 3-pyrrolidinecarboxylicacid, pyrrolidine-3-carboxylicacid, dl-pyrrolidine-3-carboxylic acid, beta-dl-proline, pubchem10439, acmc-209mcv, acmc-20a9vh
PubChem CID 3034645
IUPAC Name pyrrolidine-3-carboxylic acid
SMILES C1CNCC1C(=O)O

Specifications

Melting Point 182.0°C to 184.0°C
Elemental Analysis 50.86 to max. 53.47% min. Carbon 11.86 to max. 12.47% min. Nitrogen
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 1 g
Formula Weight 115.13
Percent Purity 98%
Chemical Name or Material Pyrrolidine-3-carboxylic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Product Code
Lot Number345092 Certificate TypeCertificate of Analysis Date02/05/2023 Product Code459110010,459110050
Lot Number342935 Certificate TypeCertificate of Analysis Date06/01/2021 Product Code459110010,459110050
compliance-icons
Product Identifier
  • Pyrrolidine-3-carboxylic acid
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pyrrolidine-3-carboxylic acid, 98% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.