Learn More
Description
Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Pyrrole-3-carboxylic acid is useful for the synthesis of cholecystokinin antagonists, benzopyran antihypertensives and azepinediones. It is utilized for the preparation of a poly(pyrrole-3-carboxylic acid) modified pencil graphite electrode and its usage in the determination of serotonin (SER) in blood serum and urine samples.
Solubility
Soluble in water.
Notes
Incompatible with strong bases, strong oxidizing agents, strong reducing agents.

Specifications
Specifications
Quantity | 1 g |
Beilstein | 108642 |
Solubility Information | Soluble in water. |
Formula Weight | 111.1 |
Percent Purity | ≥98% |
Chemical Name or Material | Pyrrole-3-carboxylic acid |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
T20I038 | Certificate of Analysis | 01/08/2022 | H55383.03,H55383.MD,H55383.06 |
Z17A021 | Certificate of Analysis | 23/11/2021 | H55383.03,H55383.MD,H55383.06 |
R28F021 | Certificate of Analysis | 23/11/2021 | H55383.03,H55383.MD,H55383.06 |
S02H048 | Certificate of Analysis | 23/11/2021 | H55383.03,H55383.MD,H55383.06 |
U05C002 | Certificate of Analysis | 23/11/2021 | H55383.03,H55383.MD,H55383.06 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.