Learn More
Tocris Bioscience™ Pyridostatin pentahydrochloride
Stabilizes G-quadruplexes; induces DNA damage and cell cycle arrest
3205.00 NOK
Specifications
CAS | 1781882-65-2 |
---|---|
Product Type | Pyridostatin pentahydrochloride |
Inhibitors | Pyridostatin pentahydrochloride |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18744719
|
Tocris Bioscience™
4763/10 |
10 mg |
3205.00 NOK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Binds and stabilizes G-quadruplexes, inducing DNA damage and cell cycle arrest (Kd = 490 nM); targets the proto-oncogene Src, reducing Src protein abundance and Src-dependent motility in human breast cancer cells. Also targets telomeric G-quadruplexes, inducing telomerase dysfunction. Activates the DNA-dependent protein kinase catalytic sunbunit (DNA-PKcs).Specifications
1781882-65-2 | |
Pyridostatin pentahydrochloride | |
Pyridostatin pentahydrochloride |
For Research Use Only