Learn More
Pyrex™ Polypropylene Pouring Rings
Ensures drip-free pouring. Insert Pyrex™ Polypropylene Pouring Ring into appropriately sized bottle neck.
Brand:  Pyrex 4516/80D
Product Code. 15797957
  • 3085.00 NOK / Pack of 10
Only null left
Estimated Shipment: 30-09-2025
Description
Ensures drip-free pouring. Insert Pyrex™ Polypropylene Pouring Ring into appropriately sized bottle neck.
  • For use with: media lab bottles, Series 1516, 1517, 1518, 1519 1520, 1523 and 1524
  • Color:clear
  • Autoclave:up to 140°C
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Clear
10/Pk.
Autoclavable (121°C)
Polypropylene
80mm
Pouring Ring
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Pyrex™ Polypropylene Pouring Rings > Size: 80mm