Learn More
Pyrex™ PBT Screwcaps
Use with reagent bottles, culture tubes and other screwtop bottles. Pyrex™ PBT Screwcaps come in a wide range of sizes for your laboratory needs. Sealing discs provide inert inner seal, resistance to moisture vapor, and are great for culture work.
Brand:  Pyrex 4503/30
 View more versions of this product
Product Code. 15985410
  • 1195.00 NOK / Pack of 20
Only null left
Estimated Shipment: 01-10-2025
Description
  • PTFE faced sealing disc for chemical resistance
  • Withstands hot air sterilization to 180deg.C
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Screw cap
PBT
30 mm
Autoclavable
Screw Cap
PTFE faced rubber insert
20/PK
PTFE faced Rubber
Culture tubes
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Pyrex™ PBT Screwcaps > Thread size: SVL 30mm