Learn More
Pyrex™ Gas Distribution Tube
Disperse gas in liquids to produce the optimum bubble size. Pyrex™ gas distribution tubes come in a variety of porosities.
741.52 NOK - 965.12 NOK
Specifications
Material | Borosilicate Glass (3.3) |
---|---|
Overall Length (Metric) | 300 mm |
For Use With (Application) | For the dispersion of gas in liquids |
Includes | Sintered thimble |
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | ||||||
15608166
|
Pyrex
3830/04M |
Porosity: 2 |
851.00 NOK
Pack of 2 |
Estimated Shipment: 14-07-2025
Log in to see stock. |
||||||
12663232
|
Pyrex
3830/02M |
Porosity: 1 |
879.84 NOK
Pack of 2 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
||||||
12547866
|
Pyrex
3830/06M |
Porosity: 3 |
741.52 NOK
Pack of 2 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
||||||
10217634
|
Pyrex
3830/00M |
Porosity: 0 |
965.12 NOK
Pack of 2 |
Estimated Shipment: 28-08-2025
Log in to see stock. |
||||||
Description
Disperse gas in liquids to produce the optimum bubble size. Pyrex™ gas distribution tubes come in a variety of porosities.
- Ideal for the dispersion of gas in liquids
- Borosilicate glass tube with sintered thimble
- Choice of thimble porosities to achieve optimum gas bubble size
Specifications
Borosilicate Glass (3.3) | |
300 mm | |
For the dispersion of gas in liquids | |
Sintered thimble |
Product Title
Pyrex™ Gas Distribution Tube