Learn More
Pyrex™ Funnel Repair Stopcocks with PTFE Key
Contains one flared end for connection to the base of the funnel
Brand: Pyrex SST704
This item is not returnable.
View return policy
Description
- Contains one flared end for connection to the base of the funnel and 100mm long stem with angled tip
- Straight bore
Specifications
Funnel Repair | |
Stopcock diameter: 18.8mm; Key bore: 4mm | |
Glass | |
PTFE key |
Product Content Correction
Product Title
Pyrex™ Funnel Repair Stopcocks with PTFE Key > Key Bore Size: 4mm