Learn More
Pyrex™ Funnel for Filtration System
For use with Pyrex™ Borosilicate Glass Filtration System.
Brand: Pyrex 5809/2
1399.42 NOK valid until 2025-07-13
Use promo code "24135" to get your promotional price.
Description
For use with Pyrex™ Borosilicate Glass Filtration System.
Specifications
Autoclavable | |
Borosilicate Glass | |
Filtration System Funnel |
Filtration System | |
1EA | |
300 mL |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%