Learn More
PYREX™ Culture Flask Closure
For use with 1135 culture flasks, 1138/10 Fernbach flasks and 1141 baffled flasks.
Brand:  Pyrex 4550/38M
 View more versions of this product
Product Code. 12687266
  • 1630.00 NOK / Pack of 10
Only null left
Estimated Shipment: 23-09-2025
Description
  • Two types of closure - Anodised aluminum - Two-position polypropylene
  • Suitable for autoclaving
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aluminum
10
Autoclavable
Documents
Product Information
Certificates
Promotions
Promotions
Product Content Correction
Product Title
PYREX™ Culture Flask Closure > 1135 culture flasks; 1138/10 Fernbach flasks