Learn More
Pyrex™ Quickfit™ Borosilicate Glass Florentine Flask
Remove solid residues with ease, using this robust, pear-shaped flask. Quickfit™ Borosilicate Glass Florentine Flask features a large surface area for rapid evaporation.
Brand:  Pyrex™ FD100/4RE
362.84 NOK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
 View more versions of this product
Product Code. 11921025
  • 403.00 NOK / Each
Estimated Shipment: 04-07-2025
to see stock.
Explore more special offers
Description
Remove solid residues with ease, using this robust, pear-shaped flask. Quickfit™ Borosilicate Glass Florentine Flask features a large surface area for rapid evaporation.
  • Ideal for use with rotary evaporators
  • Large surface area for rapid evaporation
  • Robust heavy wall construction for added strength
  • Shape allows easy removal of solid residues
TRUSTED_SUSTAINABILITY
Specifications
Specifications
100 mL
Borosilicate Glass
107 mm
29/32
Documents
Product Information
Product Title
Pyrex™ Quickfit™ Borosilicate Glass Florentine Flask > Capacity: 100mL