Learn More
Pyrex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap
Store media and cultures securely with a screw cap closure. Pryex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap resist thermal shock and chemical corrosion.
Brand:  Pyrex™ 1137/06M
 View more versions of this product
Product Code. 10029219
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
Store media and cultures securely with a screw cap closure. Pryex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap resist thermal shock and chemical corrosion.
  • PBT screw caps with PTFE faced sealing disc
  • Erlenmeyer pattern
  • Manufactured from Pyrex™ Borosilicate Glass
  • Highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
  • Graduated to full capacity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SVL™ Phenolic, PTFE-faced with rubber sealing disc
Borosilicate Glass
Approximate graduations
Yes
85 mm
250 mL
130 mm
Conical
Screw Cap
42
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products
One moment while we fetch your results.
Promotions
Promotions
Product Content Correction
Product Title
Pyrex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap > Capacity: 250mL