Learn More
Pyrex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap
Store media and cultures securely with a screw cap closure. Pryex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap resist thermal shock and chemical corrosion.
Brand:  Pyrex™ 1137/06M
 View more versions of this product
Product Code. 10029219
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
Store media and cultures securely with a screw cap closure. Pryex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap resist thermal shock and chemical corrosion.
  • PBT screw caps with PTFE faced sealing disc
  • Erlenmeyer pattern
  • Manufactured from Pyrex™ Borosilicate Glass
  • Highest resistance to thermal shock and chemical corrosion
  • White enamel graduations and marking spot
  • Graduated to full capacity
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SVL™ Phenolic, PTFE-faced with rubber sealing disc
Borosilicate Glass
Approximate graduations
Yes
85 mm
250 mL
130 mm
Conical
Screw Cap
42
Certificates
View accessory products
Product Title
Pyrex™ Borosilicate Glass Erlenmeyer Flasks with Screw Cap > Capacity: 250mL