Learn More
Lenz Laborglasintrumente Pycnometers acc. To Gay Lussac Unadjusted
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 05630013
253.56 NOK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Gay Lussac pycnometers acc. to DIN ISO 3507
- Unadjusted, stopper NS 10/19
- With capillary, stopper top ground and polished
- Imprint of nominal capacity at the bottom
- Material: borosilicate glass 3.3
Specifications
10 mL | |
Borosilicate Glass (3.3) |
Product Content Correction
Product Title
Lenz Laborglasintrumente Pycnometers acc. To Gay Lussac Unadjusted > 10 mL