Learn More
Lenz Laborglasinstrumente™ Pycnometers
694.00 NOK - 2670.00 NOK
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
10375781
|
Lenz Laborglasinstrumente™
05630213 |
10 mL |
694.00 NOK
Each |
Estimated Shipment: 22-09-2025
Log in to see stock. |
||||||
10345881
|
Lenz Laborglasinstrumente™
05640022 |
25 mL |
2460.00 NOK
Each |
Estimated Shipment: 22-09-2025
Log in to see stock. |
||||||
10080261
|
Lenz Laborglasinstrumente™
05640037 |
100 mL |
2670.00 NOK
Each |
Estimated Shipment: 22-09-2025
Log in to see stock. |
||||||
Specifications
Pycnometer | |
Pycnometer |
Product Title
Lenz Laborglasinstrumente™ Pycnometers